Anti-PARP1 (Asp214, 89 kD) cleaved Rabbit Polyclonal Antibody
CSTA9544S
- Antibody type:Primary
- Antigen symbol:PARP1 (Asp214, 89 kD) cleaved
- Clonality:Polyclonal
- Host:Rabbit
- Reactivity:Human,Mouse
- Western blot:Yes
- Seq:02
- Sequence:GTDEVAKKKSRKETDKYSC
- Storage temperature:–20 °C
- Shipping temperature:Cold
- Pk:100 µl
Type: Primary
Antigen: PARP1 (Asp214, 89 kD) cleaved
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse