Anti-PARP1 (Asp214, 89 kD) cleaved Rabbit Polyclonal Antibody
Supplier: Cell Signaling
Certificates
About this item
Type: Primary
Antigen: PARP1 (Asp214, 89 kD) cleaved
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse
Specifications
- Catalog No:
- CSTA9541L
- CSTA9541S
- Antigen symbol:
- PARP1 (Asp214, 89 kD) cleaved
- PARP1 (Asp214, 89 kD) cleaved
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Western blot:
- Yes
- Yes
- Sequence:
- GVDEVAKKKSKKEKDKDSC
- GVDEVAKKKSKKEKDKDSC
- Storage temperature:
- –20 °C
- –20 °C
- Shipping temperature:
- Cold
- Cold
Specifications
Frequently Bought Together