Anti-Smad3 (Ser423/Ser425) phosphate Rabbit Monoclonal Antibody [clone: C25A9]
Catalog # CSTA9520S
Supplier: Cell Signaling
Specifications
- Antibody type:Primary
- Antigen symbol:Smad3 (Ser423/Ser425) phosphate
- Clonality:Monoclonal
- Clone:C25A9
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Seq:02
- ChIP:Yes
- Sequence:CMGSPSIRCSsVs
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: Smad3 (Ser423/Ser425) phosphate
Clonality: Monoclonal
Clone: C25A9
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat