Anti-STAT5A (Tyr694) phosphate Rabbit Monoclonal Antibody [clone: C71E5] (Alexa Fluor 647)
Catalog # CSTA9365S
Supplier: Cell Signaling
Specifications
- Antibody type:Primary
- Antigen symbol:STAT5A (Tyr694) phosphate
- Clonality:Monoclonal
- Clone:C71E5
- Conjugation:Alexa Fluor® 647
- Flow cytometry:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Antigen synonyms:STAT5B (Tyr699) phosphate
- Sequence:CLAKAVDGyVKPQIKQ
- Storage temperature:+4 °C
- Shipping temperature:Cold
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: STAT5A (Tyr694) phosphate
Clonality: Monoclonal
Clone: C71E5
Conjugation: Alexa Fluor 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse