Anti-p38-alpha MAPK Mouse Monoclonal Antibody [clone: L53F8]
CSTA9228S
- Antibody type:Primary
- Antigen symbol:p38-alpha MAPK
- Clonality:Monoclonal
- Clone:L53F8
- Host:Mouse
- Isotype:IgG1
- Reactivity:S. cerevisiae,Pig,Monkey,Human,Rat,Mouse
- Western blot:Yes
- Storage temperature:–20 °C
- Shipping temperature:Cold
- Size:100 ul
- Pk:1 KIT
Type: Primary
Antigen: p38-alpha MAPK
Clonality: Monoclonal
Clone: L53F8
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human;Mouse;Rat;Monkey;Pig;S. cerevisiae