Anti-PARP1 (Asp214, 89 kD) cleaved Rabbit Monoclonal Antibody [clone: D64E10] (Alexa Fluor 555)
CSTA6894S
- Antibody type:Primary
- Antigen symbol:PARP1 (Asp214, 89 kD) cleaved
- Clonality:Monoclonal
- Clone:D64E10
- Conjugation:Alexa Fluor® 555
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Monkey,Human
- Sequence:GVDEVAKKKSKKEKDKDSC
- Storage temperature:+4 °C
- Shipping temperature:Cold
- Pk:100 µl
Type: Primary
Antigen: PARP1 (Asp214, 89 kD) cleaved
Clonality: Monoclonal
Clone: D64E10
Conjugation: Alexa Fluor 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Monkey