Anti-14-3-3 eta Rabbit Monoclonal Antibody [clone: D23B7]
CSTA5521S
- Antibody type:Primary
- Antigen symbol:14-3-3 eta
- Clonality:Monoclonal
- Clone:D23B7
- Host:Rabbit
- Reactivity:Pig,Monkey,Human,Bovine,Rat,Mouse
- Western blot:Yes
- Seq:02
- Sequence:KAVTELNEPLSNEDRNLLSC
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Pk:100 µl
Type: Primary
Antigen: 14-3-3 eta
Clonality: Monoclonal
Clone: D23B7
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat;Monkey;Bovine;Pig