Anti-p53 Rabbit Monoclonal Antibody [clone: 7F5] (Alexa Fluor 488)
CSTA5429S
- Antibody type:Primary
- Antigen symbol:p53
- Clonality:Monoclonal
- Clone:7F5
- Conjugation:Alexa Fluor® 488
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Monkey,Human
- Storage temperature:+4 °C
- Shipping temperature:Cold
- Pk:100 µl
Type: Primary
Antigen: p53
Clonality: Monoclonal
Clone: 7F5
Conjugation: Alexa Fluor 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Monkey