Anti-HER3/ErbB3 (Tyr1289) phosphate Rabbit Monoclonal Antibody [clone: 21D3]
Supplier: Cell Signaling
Certificates
About this item
Type: Primary
Antigen: HER3/ErbB3 (Tyr1289) phosphate
Clonality: Monoclonal
Clone: 21D3
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse
Specifications
- Catalog No:
- CSTA4791L
- CSTA4791S
- Antigen symbol:
- HER3/ErbB3 (Tyr1289) phosphate
- HER3/ErbB3 (Tyr1289) phosphate
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- 21D3
- 21D3
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Western blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- Sequence:
- CEQGyEEMRAFQ
- CEQGyEEMRAFQ
- Storage temperature:
- –20 °C
- –20 °C
- Shipping temperature:
- Cold
- Cold
Specifications
Frequently Bought Together