Anti-flotillin-2 Rabbit Monoclonal Antibody [clone: C42A3]
Catalog # CSTA3436S
Supplier: Cell Signaling
Specifications
- Antibody type:Primary
- Antigen symbol:flotillin-2
- Clonality:Monoclonal
- Clone:C42A3
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Monkey,Human,Rat,Mouse
- Western blot:Yes
- Sequence:CKKEMLDVKFMADTKIADSKR
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: flotillin-2
Clonality: Monoclonal
Clone: C42A3
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat;Monkey