Anti-PDGFR-a Rabbit Monoclonal Antibody [clone: D1E1E]
Catalog # CSTA3174S
Supplier: Cell Signaling
Specifications
- Antibody type:Primary
- Antigen symbol:PDGFR-a
- Clonality:Monoclonal
- Clone:D1E1E
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Seq:02
- Sequence:CGIDSSDLVEDSFL
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: PDGFR-a
Clonality: Monoclonal
Clone: D1E1E
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse