Anti-Vav2 Rabbit Monoclonal Antibody [clone: C64H2]
Catalog # CSTA2848S
Supplier: Cell Signaling
Specifications
- Antibody type:Primary
- Antigen symbol:Vav2
- Clonality:Monoclonal
- Clone:C64H2
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Monkey,Human
- Western blot:Yes
- Sequence:CPVDGRPPISRPPSREIDYTAYP
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: Vav2
Clonality: Monoclonal
Clone: C64H2
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Monkey