Anti-ARID1A Rabbit Monoclonal Antibody [clone: D2A8U]
Catalog # CSTA12354S
Supplier: Cell Signaling
Specifications
- Antibody type:Primary
- Antigen symbol:ARID1A
- Clonality:Monoclonal
- Clone:D2A8U
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Monkey,Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- ChIP:Yes
- Sequence:CGPYDRVRTEPGIGPEGNMSTG
- Storage temperature:–20 °C
- Shipping temperature:Cold
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: ARID1A
Clonality: Monoclonal
Clone: D2A8U
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat;Monkey