Anti-PARP1 (Asp214, 89 kD) cleaved Mouse Monoclonal Antibody [clone: 7C9]
CSTA9548S
- Antibody type:Primary
- Antigen symbol:PARP1 (Asp214, 89 kD) cleaved
- Clonality:Monoclonal
- Clone:7C9
- Host:Mouse
- Isotype:IgG2b
- Reactivity:Mouse
- Western blot:Yes
- Seq:02
- Storage temperature:–20 °C
- Shipping temperature:Cold
- Pk:100 µl
Type: Primary
Antigen: PARP1 (Asp214, 89 kD) cleaved
Clonality: Monoclonal
Clone: 7C9
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Mouse