Anti-PARP1 (Asp214, 89 kD) cleaved Mouse Monoclonal Antibody [clone: 19F4]
Supplier: Cell Signaling
Certificates
About this item
Type: Primary
Antigen: PARP1 (Asp214, 89 kD) cleaved
Clonality: Monoclonal
Clone: 19F4
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Monkey
Specifications
- Catalog No:
- CSTA9546S
- CSTA9546L
- Antigen symbol:
- PARP1 (Asp214, 89 kD) cleaved
- PARP1 (Asp214, 89 kD) cleaved
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- 19F4
- 19F4
- Reactivity:
- Monkey, Human
- Monkey, Human
- Host:
- Mouse
- Mouse
- Isotype:
- IgG1
- IgG1
- Western blot:
- Yes
- Yes
- Storage temperature:
- –20 °C
- –20 °C
- Shipping temperature:
- Cold
- Cold
Specifications
Frequently Bought Together