Anti-Smad3 (Ser423/Ser425) phosphate Rabbit Monoclonal Antibody [clone: D27F4]
Catalog # CSTA8828S
Supplier: Cell Signaling
Specifications
- Antibody type:Primary
- Antigen symbol:Smad3 (Ser423/Ser425) phosphate
- Clonality:Monoclonal
- Clone:D27F4
- Host:Rabbit
- Isotype:IgG
- Reactivity:Monkey,Human,Rat,Mouse
- Western blot:Yes
- Antigen synonyms:Smad2 (Ser465/Ser467) phosphate
- Sequence:CGSPSVRCSsMs
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: Smad3 (Ser423/Ser425) phosphate
Clonality: Monoclonal
Clone: D27F4
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Monkey