Anti-4E-BP1 (Thr37/Thr46) phosphate Rabbit Monoclonal Antibody [clone: 236B4] (PE)
CSTA7547S
- Antibody type:Primary
- Antigen symbol:4E-BP1 (Thr37/Thr46) phosphate
- Clonality:Monoclonal
- Clone:236B4
- Conjugation:PE (Phycoerythrin)
- Flow cytometry:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Drosophila melanogaster,Monkey,Human,Rat,Mouse
- Storage temperature:4 °C
- Shipping temperature:Cold
- Pk:100 µl
Type: Primary
Antigen: 4E-BP1 (Thr37/Thr46) phosphate
Clonality: Monoclonal
Clone: 236B4
Conjugation: PE
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Monkey, D. melanogaster