Order Entry
Netherlands
ContactUsLinkComponent
Anti-PARP1 (Asp214, 89 kD) cleaved Rabbit Monoclonal Antibody [clone: D64E10] (Alexa Fluor 647)
Anti-PARP1 (Asp214, 89 kD) cleaved Rabbit Monoclonal Antibody [clone: D64E10] (Alexa Fluor 647)
Catalog # CSTA6987S
Supplier:  Cell Signaling
Anti-PARP1 (Asp214, 89 kD) cleaved Rabbit Monoclonal Antibody [clone: D64E10] (Alexa Fluor 647)
Catalog # CSTA6987S
Supplier:  Cell Signaling

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    PARP1 (Asp214, 89 kD) cleaved
  • Clonality:
    Monoclonal
  • Clone:
    D64E10
  • Conjugation:
    Alexa Fluor® 647
  • Flow cytometry:
    Yes
  • Host:
    Rabbit
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Monkey,
    Human
  • Sequence:
    GVDEVAKKKSKKEKDKDSC
  • Storage temperature:
    4 °C
  • Shipping temperature:
    Cold
  • Pk:
    100 µl

Specifications

About this item

Type: Primary
Antigen: PARP1 (Asp214, 89 kD) cleaved
Clonality: Monoclonal
Clone: D64E10
Conjugation: Alexa Fluor 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Monkey