Anti-PARP1 (Asp214, 89 kD) cleaved Rabbit Monoclonal Antibody [clone: D64E10]
CSTA5625S
- Antibody type:Primary
- Antigen symbol:PARP1 (Asp214, 89 kD) cleaved
- Clonality:Monoclonal
- Clone:D64E10
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Monkey,Human
- Western blot:Yes
- Seq:02
- Sequence:GVDEVAKKKSKKEKDKDSC
- Storage temperature:–20 °C
- Shipping temperature:Cold
- Pk:100 µl
Type: Primary
Antigen: PARP1 (Asp214, 89 kD) cleaved
Clonality: Monoclonal
Clone: D64E10
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Monkey