Anti-S6 Ribosomal Protein (Ser235/Ser236) phosphate Rabbit Monoclonal Antibody [clone: D57.2.2E] (PE)
Catalog # CSTA5316S
Supplier: Cell Signaling
Specifications
- Antibody type:Primary
- Antigen symbol:S6 Ribosomal Protein (Ser235/Ser236) phosphate
- Clonality:Monoclonal
- Clone:D57.2.2E
- Conjugation:PE (Phycoerythrin)
- Flow cytometry:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:S. cerevisiae,Monkey,Human,Mink,Rat,Mouse
- Sequence:CRRLssLRASTSKSES
- Storage temperature:4 °C
- Shipping temperature:Cold
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: S6 Ribosomal Protein (Ser235/Ser236) phosphate
Clonality: Monoclonal
Clone: D57.2.2E
Conjugation: PE
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Monkey, Mink, S. cerevisiae