Anti-S6 Ribosomal Protein (Ser240/Ser244) phosphate Rabbit Monoclonal Antibody [clone: D68F8] (Alexa Fluor 647)
Catalog # CSTA5044S
Supplier: Cell Signaling
Specifications
- Antibody type:Primary
- Antigen symbol:S6 Ribosomal Protein (Ser240/Ser244) phosphate
- Clonality:Monoclonal
- Clone:D68F8
- Conjugation:Alexa Fluor® 647
- Flow cytometry:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Monkey,Human,Rat,Mouse
- Storage temperature:4 °C
- Shipping temperature:Cold
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: S6 Ribosomal Protein (Ser240/Ser244) phosphate
Clonality: Monoclonal
Clone: D68F8
Conjugation: Alexa Fluor 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Monkey