Anti-TrkA (Tyr496) phosphate Rabbit Monoclonal Antibody [clone: C35G9]
Catalog # CSTA4619S
Supplier: Cell Signaling
Specifications
- Antibody type:Primary
- Antigen symbol:TrkA (Tyr496) phosphate
- Clonality:Monoclonal
- Clone:C35G9
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human,Rat
- Western blot:Yes
- Seq:02
- Environmentally Preferable:
- Antigen synonyms:TrkB (Tyr516) phosphate
- Sequence:CGHIIENPQyFSDAGVH
- Storage temperature:–20 °C
- Shipping temperature:Cold
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: TrkA (Tyr496) phosphate
Clonality: Monoclonal
Clone: C35G9
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat