Anti-Catenin-beta (Ser675) phosphate Rabbit Monoclonal Antibody [clone: D2F1]
Catalog # CSTA4176S
Supplier: Cell Signaling
Specifications
- Antibody type:Primary
- Antigen symbol:Catenin-beta (Ser675) phosphate
- Clonality:Monoclonal
- Clone:D2F1
- Host:Rabbit
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Seq:02
- Sequence:CYKKRLsVELTS
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: Catenin-beta (Ser675) phosphate
Clonality: Monoclonal
Clone: D2F1
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat