Anti-Mer Rabbit Monoclonal Antibody [clone: D21F11] (Sepharose Bead)
Catalog # CSTA3551S
Supplier: Cell Signaling
Specifications
- Antibody type:Primary
- Antigen symbol:Mer
- Clonality:Monoclonal
- Clone:D21F11
- Conjugation:Sepharose
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human
- Environmentally Preferable:
- Sequence:CTAEVHDSKPHEGRYILNGGS
- Storage temperature:–20 °C
- Shipping temperature:Cold
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: Mer
Clonality: Monoclonal
Clone: D21F11
Conjugation: Sepharose Bead
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human