Anti-PARP1 (Asp214, 89 kD) cleaved Mouse Monoclonal Antibody [clone: E2T4K]
Catalog # CSTA32563S
Supplier: Cell Signaling
Specifications
- Antibody type:Primary
- Antigen symbol:PARP1 (Asp214, 89 kD) cleaved
- Clonality:Monoclonal
- Clone:E2T4K
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Sequence:GVDEVAKKKSKKEKDKDSC
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: PARP1 (Asp214, 89 kD) cleaved
Clonality: Monoclonal
Clone: E2T4K
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human