Anti-14-3-3 Binding motif [R-X-(Y/F)-X-S] phosphate Rabbit Polyclonal Antibody
Catalog # CSTA2981S
Supplier: Cell Signaling
Specifications
- Antibody type:Primary
- Antigen symbol:14-3-3 Binding motif [R-X-(Y/F)-X-S] phosphate
- Clonality:Polyclonal
- ELISA:Yes
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Reactivity:Multiple
- Western blot:Yes
- Sequence:XXXRX(F/Y)XsXPXXXC
- Storage temperature:–20 °C
- Shipping temperature:Cold
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: 14-3-3 Binding motif [R-X-(Y/F)-X-S] phosphate
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Any species