Anti-IkB-a (Ser32) phosphate Rabbit Monoclonal Antibody [clone: 14D4]
Type: Primary
Antigen: IkB-a (Ser32) phosphate
Clonality: Monoclonal
Clone: 14D4
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Monkey
- Catalog No:
- CSTA2859L
- CSTA2859S
- Antigen symbol:
- IkB-a (Ser32) phosphate
- IkB-a (Ser32) phosphate
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- 14D4
- 14D4
- Reactivity:
- Monkey, Human, Rat, Mouse
- Monkey, Human, Rat, Mouse
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- Sequence:
- LDDRHDsGLDSMKC
- LDDRHDsGLDSMKC
- Storage temperature:
- –20 °C
- –20 °C
- Shipping temperature:
- Cold
- Cold
Frequently Bought Together