Anti-VEGFR2 (Tyr1214) phosphate Rabbit Monoclonal Antibody [clone: 11A3]
Catalog # CSTA2477S
Supplier: Cell Signaling
Specifications
- Antibody type:Primary
- Antigen symbol:VEGFR2 (Tyr1214) phosphate
- Clonality:Monoclonal
- Clone:11A3
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Sequence:CDPKFHyDNTAGIS
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: VEGFR2 (Tyr1214) phosphate
Clonality: Monoclonal
Clone: 11A3
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse