Anti-p38-delta MAPK Rabbit Monoclonal Antibody [clone: 10A8]
Catalog # CSTA2308S
Supplier: Cell Signaling
Specifications
- Antibody type:Primary
- Antigen symbol:p38-delta MAPK
- Clonality:Monoclonal
- Clone:10A8
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human,Rat
- Western blot:Yes
- Seq:02
- Sequence:CIARKDSRRRSGMKL
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: p38-delta MAPK
Clonality: Monoclonal
Clone: 10A8
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat