Anti-H3 (Arg2) methylate Rabbit Polyclonal Antibody
Catalog # CSTA9707S
Supplier: Cell Signaling
Specifications
- Antibody type:Primary
- Antigen symbol:H3 (Arg2) methylate
- Clonality:Polyclonal
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Sequence:CA[R-Me2]TKQTARKST
- Storage temperature:–20 °C
- Shipping temperature:Cold
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: H3 (Arg2) methylate
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human