Anti-IkB-a Mouse Monoclonal Antibody [clone: 112B2]
Catalog # CSTA9247S
Supplier: Cell Signaling
Specifications
- Antibody type:Primary
- Antigen symbol:IkB-a
- Clonality:Monoclonal
- Clone:112B2
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG2a
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Sequence:EFTEDELPYDDCVFGGQRLTL
- Storage temperature:–20 °C
- Shipping temperature:Cold
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: IkB-a
Clonality: Monoclonal
Clone: 112B2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse, Rat