Anti-FAK (Tyr397) phosphate Rabbit Monoclonal Antibody [clone: D20B1]
CSTA8556S
- Type antilichaam:Primair
- antigeen symbool:FAK (Tyr397) phosphate
- Clonality:Monoklonaal
- Clone:D20B1
- Host:Rabbit
- ImmunoNeerslag:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Seq:02
- Environmentally Preferable:
- Sequentie:CSVSETDDyAEIIDKK
- Opslagtemperatuur:–20 °C
- Shipping temperature:Ambient
- VPE:100 µl
Type: Primary
Antigen: FAK (Tyr397) phosphate
Clonality: Monoclonal
Clone: D20B1
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human