Anti-STAT3 (Tyr705) phosphate Rabbit Monoclonal Antibody [clone: D3A7] (PE)
CSTA8119S
- Antibody type:Primary
- Antigen symbol:STAT3 (Tyr705) phosphate
- Clonality:Monoclonal
- Clone:D3A7
- Conjugation:PE (Phycoerythrin)
- Flow cytometry:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Monkey,Human,Rat,Mouse
- Sequence:ADPGSAAPyLKTKFIC
- Storage temperature:+4 °C
- Shipping temperature:Cold
- Pk:100 µl
Type: Primary
Antigen: STAT3 (Tyr705) phosphate
Clonality: Monoclonal
Clone: D3A7
Conjugation: PE
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Monkey