Anti-PDCD8 Rabbit Monoclonal Antibody [clone: D39D2] (Sepharose Bead)
CSTA5939S
- Antibody type:Primary
- Antigen symbol:PDCD8
- Clonality:Monoclonal
- Clone:D39D2
- Conjugation:Sepharose
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Monkey,Human,Rat,Mouse
- Environmentally Preferable:
- Sequence:CDNPKSATEQSGTGIRSES
- Storage temperature:–20 °C
- Shipping temperature:Cold
- Pk:400 µl
Type: Primary
Antigen: PDCD8
Clonality: Monoclonal
Clone: D39D2
Conjugation: Sepharose Bead
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Monkey