Anti-STEP (Ser245) non-phosphate Rabbit Monoclonal Antibody [clone: D74H3]
CSTA5659S
- Antibody type:Primary
- Antigen symbol:STEP (Ser245) non-phosphate
- Clonality:Monoclonal
- Clone:D74H3
- Host:Rabbit
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Sequence:CGLQERRGSNVSLT
- Storage temperature:–20 °C
- Shipping temperature:Cold
- Pk:100 µl
Type: Primary
Antigen: STEP (Ser245) non-phosphate
Clonality: Monoclonal
Clone: D74H3
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat