Anti-ARHGAP42 (Tyr376) phosphate Rabbit Monoclonal Antibody [clone: D45E9]
Catalog # CSTA5617S
Supplier: Cell Signaling
Specifications
- Antibody type:Primary
- Antigen symbol:ARHGAP42 (Tyr376) phosphate
- Clonality:Monoclonal
- Clone:D45E9
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Monkey,Human,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Sequence:KEPIyTLPAIISKKC
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: ARHGAP42 (Tyr376) phosphate
Clonality: Monoclonal
Clone: D45E9
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Monkey