Anti-S6 Ribosomal Protein (Ser235/Ser236) phosphate Rabbit Monoclonal Antibody [clone: D57.2.2E]
Leverancier: Cell Signaling
Certificaten
Over dit artikel
Type: Primary
Antigen: S6 Ribosomal Protein (Ser235/Ser236) phosphate
Clonality: Monoclonal
Clone: D57.2.2E
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Monkey, Mink, S. cerevisiae
Specificaties
- Catalog No:
- CSTA4858S
- CSTA4858L
- Antigen symbol:
- S6 Ribosomal Protein (Ser235/Ser236) phosphate
- S6 Ribosomal Protein (Ser235/Ser236) phosphate
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- D57.2.2E
- D57.2.2E
- Reactivity:
- S. cerevisiae, Monkey, Human, Mink, Rat, Mouse
- S. cerevisiae, Monkey, Human, Mink, Rat, Mouse
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Flow cytometry:
- Yes
- Yes
- Sequence:
- CRRLssLRASTSKSES
- CRRLssLRASTSKSES
- Storage temperature:
- –20 °C
- –20 °C
- Shipping temperature:
- Ambient
- Ambient
Specificaties
Frequently Bought Together