Anti-H3 (Lys27) trimethylate Rabbit Monoclonal Antibody [clone: C36B11] (Biotinylated)
Catalog # CSTA4395S
Supplier: Cell Signaling
Specifications
- Antibody type:Primary
- Antigen symbol:H3 (Lys27) trimethylate
- Clonality:Monoclonal
- Clone:C36B11
- Conjugation:Biotin
- Host:Rabbit
- Isotype:IgG
- Reactivity:Monkey,Human,Rat,Mouse
- Western blot:Yes
- Sequence:TKAAR(K-tri-methyl)SAPATGG(TKAAR(K-tri-methyl)SAPATGG)KC
- Storage temperature:–20 °C
- Shipping temperature:Cold
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: H3 (Lys27) trimethylate
Clonality: Monoclonal
Clone: C36B11
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Monkey