Anti-RSK2 (Ser227) phosphate Rabbit Monoclonal Antibody [clone: D53A11]
Catalog # CSTA3556S
Supplier: Cell Signaling
Specifications
- Antibody type:Primary
- Antigen symbol:RSK2 (Ser227) phosphate
- Clonality:Monoclonal
- Clone:D53A11
- Host:Rabbit
- Isotype:IgG
- Reactivity:Monkey,Human,Rat,Mouse
- Western blot:Yes
- Antigen synonyms:RSK1 (Ser221) phosphate
- Sequence:HEKKAYsFAGTVEC
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: RSK2 (Ser227) phosphate
Clonality: Monoclonal
Clone: D53A11
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Monkey