Order Entry
Netherlands
ContactUsLinkComponent
Anti-PP2C alpha Rabbit Monoclonal Antibody [clone: D18C10]
Anti-PP2C alpha Rabbit Monoclonal Antibody [clone: D18C10]
Catalog # CSTA3549S
Supplier:  Cell Signaling
Anti-PP2C alpha Rabbit Monoclonal Antibody [clone: D18C10]
Catalog # CSTA3549S
Supplier:  Cell Signaling

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    PP2C alpha
  • Clonality:
    Monoclonal
  • Clone:
    D18C10
  • Flow cytometry:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Monkey,
    Human
  • Western blot:
    Yes
  • Seq:
    02
  • Sequence:
    CNRLNPYKNDDTDSTSTDDMW
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    Ambient
  • Pk:
    100 µl

Specifications

About this item

Type: Primary
Antigen: PP2C alpha
Clonality: Monoclonal
Clone: D18C10
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Monkey