Anti-ERK1 (Thr202/Tyr204) phosphate Rabbit Monoclonal Antibody [clone: D13.14.4E] (Sepharose Bead)
CSTA3510S
- Antibody type:Primary
- Antigen symbol:ERK1 (Thr202/Tyr204) phosphate
- Clonality:Monoclonal
- Clone:D13.14.4E
- Conjugation:Sepharose
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Zebrafish,Mouse,S. cerevisiae,Hamster,Pig,Monkey,Human,Bovine,Dog,Mink,D. melanogasterm,Rat
- Antigen synonyms:ERK2 (Thr185/Tyr187) phosphate
- Sequence:TGFLtEyVATRWYRC
- Storage temperature:–20 °C
- Shipping temperature:Cold
- Pk:400 µl
Type: Primary
Antigen: ERK1 (Thr202/Tyr204) phosphate
Clonality: Monoclonal
Clone: D13.14.4E
Conjugation: Sepharose Bead
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Hamster, Monkey, Mink, D. melanogasterm, Zebrafish, Bovine, Dog, Pig, S. cerevisiae