Anti-TrkC Rabbit Monoclonal Antibody [clone: C44H5]
Catalog # CSTA3376S
Supplier: Cell Signaling
Specifications
- Antibody type:Primary
- Antigen symbol:TrkC
- Clonality:Monoclonal
- Clone:C44H5
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Sequence:CRRPDDGNLFPLLEGQDSGNSNG
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: TrkC
Clonality: Monoclonal
Clone: C44H5
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat