Anti-TAB1 Rabbit Monoclonal Antibody [clone: C25E9]
Catalog # CSTA3226S
Supplier: Cell Signaling
Specifications
- Antibody type:Primary
- Antigen symbol:TAB1
- Clonality:Monoclonal
- Clone:C25E9
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Monkey,Human
- Western blot:Yes
- Sequence:CPAAGGRVYPVSVPYSSAQSTSKTS
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: TAB1
Clonality: Monoclonal
Clone: C25E9
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Monkey