Anti-PDGFR-b (Tyr1009) phosphate Rabbit Monoclonal Antibody [clone: 42F9]
Catalog # CSTA3124S
Supplier: Cell Signaling
Specifications
- Antibody type:Primary
- Antigen symbol:PDGFR-b (Tyr1009) phosphate
- Clonality:Monoclonal
- Clone:42F9
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Reactivity:Human,Mouse
- Western blot:Yes
- Seq:02
- Sequence:VLyTAVQPNEGDNDC
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: PDGFR-b (Tyr1009) phosphate
Clonality: Monoclonal
Clone: 42F9
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse