Anti-EEF1A Rabbit Polyclonal Antibody
Catalog # CSTA2551S
Supplier: Cell Signaling
Specifications
- Antibody type:Primary
- Antigen symbol:EEF1A
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoFluorescence:Yes
- Reactivity:Monkey,Human,Rat,Mouse
- Western blot:Yes
- Sequence:CVESFSDYPPLGRFAVRDMRQT
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: EEF1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat, Monkey