Anti-Synapsin-1 Rabbit Polyclonal Antibody
CSTA2312S
- Antibody type:Primary
- Antigen symbol:Synapsin-1
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:
- Reactivity:Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Sequence:CYLRRRLSDSNF
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Pk:100 µl
Type: Primary
Antigen: Synapsin-1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse, Rat