Anti-Motif (R) a-dimethyl Rabbit Monoclonal Antibody [clone: Asymmetric Di-Methyl Arginine Motif [adme-R] MultiMab™ Rabbit mAb mix]
Catalog # CSTA13522S
Supplier: Cell Signaling
Specifications
- Antibody type:Primary
- Antigen symbol:Motif (R) a-dimethyl
- Clonality:Monoclonal
- Clone:Asymmetric Di-Methyl Arginine Motif [adme-R] MultiMab™ Rabbit mAb mix
- Host:Rabbit
- Isotype:IgG
- Reactivity:Multiple
- Western blot:Yes
- Sequence:CXXXXrXXrXXXXrXXXrXXXX
- Storage temperature:–20 °C
- Shipping temperature:Cold
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: Motif (R) a-dimethyl
Clonality: Monoclonal
Clone: Asymmetric Di-Methyl Arginine Motif [adme-R] MultiMab™ Rabbit mAb mix
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Any species