Anti-PARP1 (Asp214, 89 kD) cleaved Rabbit Polyclonal Antibody
Catalog # CSTA9545S
Supplier: Cell Signaling
Specifications
- Antibody type:Primary
- Antigen symbol:PARP1 (Asp214, 89 kD) cleaved
- Clonality:Polyclonal
- Host:Rabbit
- Reactivity:Rat
- Western blot:Yes
- Seq:02
- Sequence:GIDEVAKKKSKKGKDKESC
- Storage temperature:−20 °C
- Shipping temperature:Ambient
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Type: Primary Antibodies
Antigen: PARP1 (Asp214, 89 kD) cleaved
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Hoat: Rabbit
Isotype:
Reactivity: Rat