Anti-IkB-a (Ser32/Ser36) phosphate Mouse Monoclonal Antibody [clone: '5A5]
Supplier: Cell Signaling
Certificates
About this item
Type: Primary Antibodies
Antigen: IkB-a (Ser32/Ser36) phosphate
Clonality: Monoclonal
Clone: '5A5
Conjugation:
Epitope:
Hoat: Mouse
Isotype: IgG1
Reactivity: Human,Mouse,Rat,Monkey
Specifications
- Catalog No:
- CSTA9246L
- CSTA9246S
- Antigen symbol:
- IkB-a (Ser32/Ser36) phosphate
- IkB-a (Ser32/Ser36) phosphate
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- 5A5
- 5A5
- Reactivity:
- Monkey, Human, Rat, Mouse
- Monkey, Human, Rat, Mouse
- Host:
- Mouse
- Mouse
- Isotype:
- IgG1
- IgG1
- Western blot:
- Yes
- Yes
- Sequence:
- LDDRHDsGLDsMKDC
- LDDRHDsGLDsMKDC
- Storage temperature:
- −20 °C
- −20 °C
- Shipping temperature:
- Ambient
- Ambient
Specifications
Frequently Bought Together