Anti-MKK4 (Thr261) phosphate Rabbit Polyclonal Antibody
CSTA9151S
- Antibody type:Primary
- Antigen symbol:MKK4 (Thr261) phosphate
- Clonality:Polyclonal
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Sequence:SIAKtRDAC
- Storage temperature:−20 °C
- Shipping temperature:Ambient
- Size:100 µl
- Pk:100 µl
Type: Primary Antibodies
Antigen: MKK4 (Thr261) phosphate
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Hoat: Rabbit
Isotype:
Reactivity: Human,Mouse,Rat